Thermo Scientific Chemicals

4-Aminoantipyrine, 97%, Thermo Scientific Chemicals

Catalog number: A13846.22
100 g, Each
Thermo Scientific Chemicals

4-Aminoantipyrine, 97%, Thermo Scientific Chemicals

Catalog number: A13846.22
100 g, Each
Quantity
Catalog number: A13846.22
also known as A13846-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
83-07-8
IUPAC Name
4-amino-1,5-dimethyl-2-phenyl-2,3-dihydro-1H-pyrazol-3-one
Molecular Formula
C11H13N3O
InChI Key
RLFWWDJHLFCNIJ-UHFFFAOYSA-N
SMILES
CN1N(C(=O)C(N)=C1C)C1=CC=CC=C1
Appearance (Color)
Yellow to orange to pale brown
Form
Crystals or powder or crystalline powder
Assay (HPLC)
≥96.0%
Assay (Non-aqueous acid-base Titration)
≥96.0 to ≤104.0%
Identification (FTIR)
Conforms

Description

4-Aminoantipyrine shows both analgesic and anti-inflammatory properties. It is used as a reagent for glucose determination in the presence of peroxidase and phenol. It is used as a drug dipyrone.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Aminoantipyrine shows both analgesic and anti-inflammatory properties. It is used as a reagent for glucose determination in the presence of peroxidase and phenol. It is used as a drug dipyrone.

Solubility
Soluble in water. 500 g/L (20°C), methanol. Partially soluble in diethyl ether.

Notes
Incompatible with Strong oxidizing agents, strong acids, acid chlorides, acid anhydrides.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text