Thermo Scientific Chemicals

1-Fluoro-2-nitrobenzene, 99%, Thermo Scientific Chemicals

Catalog number: A13852.30
250 g, Each
Thermo Scientific Chemicals

1-Fluoro-2-nitrobenzene, 99%, Thermo Scientific Chemicals

Catalog number: A13852.30
250 g, Each
Quantity
Catalog number: A13852.30
also known as A13852-30
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1493-27-2
IUPAC Name
1-fluoro-2-nitrobenzene
Molecular Formula
C6H4FNO2
InChI Key
PWKNBLFSJAVFAB-UHFFFAOYSA-N
SMILES
[O-][N+](=O)C1=CC=CC=C1F
Appearance (Color)
Clear yellow to orange to brown
Identification (FTIR)
Conforms
Assay (GC)
≥98.5%
Refractive Index
1.5300-1.5330 @ 20?C
Form
Liquid

Description

1-Fluoro-2-nitrobenzene is an intermediate in the synthetic preparations of various pharmaceutical goods.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Fluoro-2-nitrobenzene is an intermediate in the synthetic preparations of various pharmaceutical goods.

Solubility
Immiscible in water.

Notes
Avoid excessive heat and light.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text