Thermo Scientific Chemicals

n-Eicosane, 99%, Thermo Scientific Chemicals

Catalog number: A13853.22
100 g, Each
Thermo Scientific Chemicals

n-Eicosane, 99%, Thermo Scientific Chemicals

Catalog number: A13853.22
100 g, Each
Quantity
Catalog number: A13853.22
also known as A13853-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
112-95-8
IUPAC Name
icosane
Molecular Formula
C20H42
InChI Key
CBFCDTFDPHXCNY-UHFFFAOYSA-N
SMILES
CCCCCCCCCCCCCCCCCCCC
Assay (GC)
≥98.5% (UK-sourced material)
Appearance (Color)
White to pale yellow
Assay from Supplier's CofA
≥98.5% (US-sourced material)
Identification (FTIR)
Conforms (UK-sourced material)
Form
Crystals or powder or crystalline powder or fused solid or flakes

Description

n-Eicosane is a saturated aliphatic hydrocarbon, which is used to form candles and paraffin waxes with solar energy storage capacity. It is also used in cosmetics, lubricants, plasticizers and in the petrochemical industry. Further, it is useful for special surfactants and in the preparation of certain organic chemicals. In addition to this, it is used as a chromatographic analysis reference material.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
n-Eicosane is a saturated aliphatic hydrocarbon, which is used to form candles and paraffin waxes with solar energy storage capacity. It is also used in cosmetics, lubricants, plasticizers and in the petrochemical industry. Further, it is useful for special surfactants and in the preparation of certain organic chemicals. In addition to this, it is used as a chromatographic analysis reference material.

Solubility
Soluble in ether, petroleum ether and benzene. Slightly soluble in acetone and chloroform. Insoluble in water.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text