Thermo Scientific Chemicals

Tritylamine, 98%, Thermo Scientific Chemicals

Catalog number: A13864.14
25 g, Each
Thermo Scientific Chemicals

Tritylamine, 98%, Thermo Scientific Chemicals

Catalog number: A13864.14
25 g, Each
Quantity
Catalog number: A13864.14
also known as A13864-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
5824-40-8
IUPAC Name
triphenylmethanamine
Molecular Formula
C19H17N
InChI Key
BZVJOYBTLHNRDW-UHFFFAOYSA-N
SMILES
NC(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
Form
Crystals or powder or crystalline powder
Melting Point (clear melt)
100.0-106.0?C
Appearance (Color)
White to cream
Assay (Non-aqueous acid-base Titration)
≥97.5 to ≤102.5%

Description

Triphenylmethylamine was used to prepare diamondoid porous organic salt.1 It was also used in preparation of N-tritylated β-amino alcohols, useful building blocks in organic synthesis. It is used in the preparation of trityl- cysteine analogs as inhibitors of human mitotic kinesin.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Triphenylmethylamine was used to prepare diamondoid porous organic salt.1 It was also used in preparation of N-tritylated β-amino alcohols, useful building blocks in organic synthesis. It is used in the preparation of trityl- cysteine analogs as inhibitors of human mitotic kinesin.

Solubility
Soluble in chloroform, methanol. Insoluble in water.

Notes
Air sensitive. Store in a cool, dry, well-ventilated area. Store away from oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text