Thermo Scientific Chemicals

3,3',5,5'-Tetramethylbenzidine, 98%, Thermo Scientific Chemicals

Catalog number: A13868.03
1 g, Each
Thermo Scientific Chemicals

3,3',5,5'-Tetramethylbenzidine, 98%, Thermo Scientific Chemicals

Catalog number: A13868.03
1 g, Each
Quantity
Catalog number: A13868.03
also known as A13868-03
Price (USD)
Have Questions?

Chemical Identifiers

CAS
54827-17-7
IUPAC Name
3,3',5,5'-tetramethyl-[1,1'-biphenyl]-4,4'-diamine
Molecular Formula
C16H20N2
InChI Key
UAIUNKRWKOVEES-UHFFFAOYSA-N
SMILES
CC1=CC(=CC(C)=C1N)C1=CC(C)=C(N)C(C)=C1
Melting Point (clear melt)
166.0-172.0?C
Form
Powder
Appearance (Color)
White to cream to pale brown
Assay (GC)
≥97.5%

Description

Excellent colorimetric substrate for detection of horseradish peroxidase labelled probes; particularly useful in ELISA techniques, blue result can be read spectrophotometrically at 370 or 650 nm3,3',5,5'-Tetramethylbenzidine is used as a clinical reagent for testing blood. It is also a chromogenic substrate which is used in staining procedures in immunohistochemistry. Further, it is used as a visualizing reagent in enzyme-linked immunosorbent assays. In addition to this, it is used as a replacement for carcinogenic compounds such as benzidine and o-phenylenediamine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description
• 3,3',5,5'-Tetramethylbenzidine, also known as 4,4'-Diamino-3,3',5,5'-tetramethylbiphenyl or TMB, is a colorimetric substrate for detection of horseradish peroxidase-labeled probes
• Oxidation of 3,3',5,5'-Tetramethylbenzidine results in a pale blue end product that can be detected at 370 or 655 nm. Sulfuric acid (H2SO4) is used to stop the oxidation reaction and stabilize the reading, resulting in a yellow color that can be read at 450 nm
Application
• 3,3',5,5'-Tetramethylbenzidine is used as a chromogenic visualization agent in enzyme-linked immunosorbent assays (ELISA) or immunohistochemistry (IHC) techniques
• 3,3',5,5'-Tetramethylbenzidine has been used to detect molecules in aqueous samples such as copper ion and hypochlorite
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text