Thermo Scientific Chemicals

trans-4-Methoxycinnamic acid, 98%, Thermo Scientific Chemicals

Catalog number: A13870.30
250 g, Each
Thermo Scientific Chemicals

trans-4-Methoxycinnamic acid, 98%, Thermo Scientific Chemicals

Catalog number: A13870.30
250 g, Each
Quantity
Catalog number: A13870.30
also known as A13870-30
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
943-89-5
IUPAC Name
(2E)-3-(4-methoxyphenyl)prop-2-enoic acid
Molecular Formula
C10H10O3
InChI Key
AFDXODALSZRGIH-QPJJXVBHSA-N
SMILES
COC1=CC=C(C=CC(O)=O)C=C1
Identification (FTIR)
Conforms
Form
Crystals or powder or crystalline powder
Assay (Silylated GC)
≥97.5%
Melting Point
170.0-176.0?C
Appearance (Color)
White to cream

Description

Esters derived from trans-4-methoxycinnamic acid are effective absorbers of UV radiation. The starting material was trans-4-methoxycinnamic acid and 3,5dimethoxybenzoic acid methyl ester in isolated avenanthramide alkaloids synthsis. trans-4-Methoxycinnamic acid was converted into its acid chloride (yield 98%) with thionyl chloride. Employed in organic synthesis of pharmaceutical intermediates, synthetic anti-adrenergic drugs esmolol, cosmetics ultraviolet absorption.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Esters derived from trans-4-methoxycinnamic acid are effective absorbers of UV radiation. The starting material was trans-4-methoxycinnamic acid and 3,5dimethoxybenzoic acid methyl ester in isolated avenanthramide alkaloids synthsis. trans-4-Methoxycinnamic acid was converted into its acid chloride (yield 98%) with thionyl chloride. Employed in organic synthesis of pharmaceutical intermediates, synthetic anti-adrenergic drugs esmolol, cosmetics ultraviolet absorption.

Solubility
Soluble in ethanol, ethyl acetate and other organic solvents.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text