Thermo Scientific Chemicals

Pyruvic acid, 98%, Thermo Scientific Chemicals

Catalog number: A13875.0E
2500 g, Each
Thermo Scientific Chemicals

Pyruvic acid, 98%, Thermo Scientific Chemicals

Catalog number: A13875.0E
2500 g, Each
Quantity
Catalog number: A13875.0E
also known as A13875-0E
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
127-17-3
IUPAC Name
2-oxopropanoic acid
Molecular Formula
C3H4O3
InChI Key
LCTONWCANYUPML-UHFFFAOYSA-N
SMILES
CC(=O)C(O)=O
Appearance (Color)
Clear colorless to yellow
Comment
Material sourced in the U.S. and in other countries
Identification (FTIR)
Conforms (non-U.S. sourced material)
Refractive Index
1.4260-1.4320 @ 20°C (non-U.S. sourced material)
Assay (Aqueous acid-base Titration)
≥97.5 to ≤102.5% (non-U.S. sourced material)

Description

Pyruvic acid is used as a component in culture broths and media as commercial red seaweed polysaccharide. It is involved in the construction of amino acid alanine as well as supplies energy to living cells via citric acid cycle (Krebs cycle). It reacts with N-acetyl mannosamine by an aldol-type condensation to prepare sialic acid. It is employed to study the cultivation of soil bacteria as micro colonies using soil substrate membrane system. It finds application in liquid chromatography and in the determination of organic acids in red wine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Pyruvic acid is used as a component in culture broths and media as commercial red seaweed polysaccharide. It is involved in the construction of amino acid alanine as well as supplies energy to living cells via citric acid cycle (Krebs cycle). It reacts with N-acetyl mannosamine by an aldol-type condensation to prepare sialic acid. It is employed to study the cultivation of soil bacteria as micro colonies using soil substrate membrane system. It finds application in liquid chromatography and in the determination of organic acids in red wine.

Solubility
Miscible with chloroform and methanol.

Notes
Store in cool place. Incompatible with strong oxidizing agents, strong bases and reducing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text