4'-Nitroacetophenone, 98%, Thermo Scientific Chemicals
4'-Nitroacetophenone, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4'-Nitroacetophenone, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
Catalog number A13877.18
also known as A13877-18
Price (USD)/ Each
26.65
Online exclusive
29.80 
Save 3.15 (11%)
-
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (USD)/ Each
26.65
Online exclusive
29.80 
Save 3.15 (11%)
Add to cart
4'-Nitroacetophenone, 98%, Thermo Scientific Chemicals
Catalog numberA13877.18
Price (USD)/ Each
26.65
Online exclusive
29.80 
Save 3.15 (11%)
-
Add to cart
Chemical Identifiers
CAS100-19-6
IUPAC Name1-(4-nitrophenyl)ethan-1-one
Molecular FormulaC8H7NO3
InChI KeyYQYGPGKTNQNXMH-UHFFFAOYSA-N
SMILESCC(=O)C1=CC=C(C=C1)[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Assay (GC)≥97.5%
Appearance (Color)Yellow
Identification (FTIR)Conforms
Melting Point (clear melt)76.0-83.0?C
4'-Nitroacetophenone is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4′-Nitroacetophenone is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals.

Solubility
Insoluble in water.

Notes
Store in a cool, dry, well-ventilated area. Store away from oxidizing agent, strong alkalis.
RUO – Research Use Only

General References:

  1. Joel M. Hawkins and Teresa W. Makowski. Optimizing Selective Partial Hydrogenations of 4-Nitroacetophenone via Parallel Reaction Screening. Org. Proc. Res. Dev. 2001, 5 (3), 328-330.
  2. Gerald M. Cohen & Ian R. Flockhart. The Partial Purification and Properties of a Human Erythrocyte 4-Nitroacetophenone Reductase. Xenobiotica: the fate of foreign compounds in biological systems. 1975, 5 (4), 4993-4996.