4'-Nitroacetophenone is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4′-Nitroacetophenone is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals.
Solubility
Insoluble in water.
Notes
Store in a cool, dry, well-ventilated area. Store away from oxidizing agent, strong alkalis.
RUO – Research Use Only
General References:
- Joel M. Hawkins and Teresa W. Makowski. Optimizing Selective Partial Hydrogenations of 4-Nitroacetophenone via Parallel Reaction Screening. Org. Proc. Res. Dev. 2001, 5 (3), 328-330.
- Gerald M. Cohen & Ian R. Flockhart. The Partial Purification and Properties of a Human Erythrocyte 4-Nitroacetophenone Reductase. Xenobiotica: the fate of foreign compounds in biological systems. 1975, 5 (4), 4993-4996.