1,2-Dichlorobenzene, 99%, Thermo Scientific Chemicals
1,2-Dichlorobenzene, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1,2-Dichlorobenzene, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
10,000 g
500 g
2500 g
Catalog number A13881.0C
also known as A13881-0C
Price (USD)/ Each
442.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
520.00 
Save 77.35 (15%)
-
Add to cart
Quantity:
10,000 g
Request bulk or custom format
Price (USD)/ Each
442.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
520.00 
Save 77.35 (15%)
Add to cart
1,2-Dichlorobenzene, 99%, Thermo Scientific Chemicals
Catalog numberA13881.0C
Price (USD)/ Each
442.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
520.00 
Save 77.35 (15%)
-
Add to cart
Chemical Identifiers
CAS95-50-1
IUPAC Name1,2-dichlorobenzene
Molecular FormulaC6H4Cl2
InChI KeyRFFLAFLAYFXFSW-UHFFFAOYSA-N
SMILESClC1=CC=CC=C1Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive Index1.5500-1.5530 @ 20°C
Appearance (Color)Clear colorless
Assay from Suppliers CofA≥98.5% (U.S. specification)
Identification (FTIR)Conforms (non-U.S. specification)
CommentSpecification differs for U.S. and non-U.S. material where indicated
View more
1,2-Dichlorobenzene is widely used as a precursor to prepare 1,2-dichloro-4-nitrobenzene, which is an intermediate in the synthesis of agrochemicals. It acts as a solvent for fullerenes. It is also used in softening and removing carbon-based contamination on metal surfaces. It also used as food preservative, motor oil additive, lubricant and paint.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,2-Dichlorobenzene is widely used as a precursor to prepare 1,2-dichloro-4-nitrobenzene, which is an intermediate in the synthesis of agrochemicals. It acts as a solvent for fullerenes. It is also used in softening and removing carbon-based contamination on metal surfaces. It also used as food preservative, motor oil additive, lubricant and paint.

Solubility
Miscible with water and most of the organic solvents.

Notes
Keep away from sources of ignition. Incompatible with strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Chen, N. Y.; Yang, S. C.; Liu, M. C.; Lee, J. F.; Chang, J. R. Pellet Vanadia Catalysts for Oxidative Destruction of 1,2-Dichlorobenzene: Roles of the Grafted TiO2 in Vanadia Morphology and Catalytic Reaction. Catal. Surv. Asia 2015, 19 (1), 38-56.
  2. Cai, T.; Huang, H.; Deng, W.; Dai, Q.; Liu, W.; Wang, X. Catalytic combustion of 1, 2-dichlorobenzene at low temperature over Mn-modified Co3O4 catalysts. Appl. Catal., B 2015, 166-167, 393-405.