Thermo Scientific Chemicals

Decahydronaphthalene, cis + trans, 98%, Thermo Scientific Chemicals

Catalog number: A13883.AP
500 mL, Each
Thermo Scientific Chemicals

Decahydronaphthalene, cis + trans, 98%, Thermo Scientific Chemicals

Catalog number: A13883.AP
500 mL, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: A13883.AP
also known as A13883-AP
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
91-17-8
IUPAC Name
decahydronaphthalene
Molecular Formula
C10H18
InChI Key
NNBZCPXTIHJBJL-UHFFFAOYSA-N
SMILES
C1CCC2CCCCC2C1
Form
Liquid
Refractive Index
1.4720-1.4760 @ 20?C
Assay (GC)
≥97.5%
Appearance (Color)
Clear colorless
Identification (FTIR)
Conforms

Description

Decahydronaphthalene is widely used as an industrial solvent for resins and fuel additives. It is a substitute for turpentine in lacquers, shoe polishes and waxes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Decahydronaphthalene is widely used as an industrial solvent for resins and fuel additives. It is a substitute for turpentine in lacquers, shoe polishes and waxes.

Solubility
Miscible with acetone, benzene, ether, methanol, aniline, decalin, chloroform, butanol and ethanol. Immiscible with water.

Notes
Store in cool place. Air, light and heat sensitive. Heat and light accelerate peroxide formation. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text