Thermo Scientific Chemicals

3-Sulfolene, 98%, Thermo Scientific Chemicals

Catalog number: A13887.36
500 g, Each
Thermo Scientific Chemicals

3-Sulfolene, 98%, Thermo Scientific Chemicals

Catalog number: A13887.36
500 g, Each
Quantity
Catalog number: A13887.36
also known as A13887-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
77-79-2
IUPAC Name
2,5-dihydro-1λ⁶-thiophene-1,1-dione
Molecular Formula
C4H6O2S
InChI Key
MBDNRNMVTZADMQ-UHFFFAOYSA-N
SMILES
O=S1(=O)CC=CC1
Appearance (Color)
White to cream
Form
Crystals or powder or crystalline powder or lumps or chunks
Melting Point (clear melt)
62.0-68.0?C
Assay (GC)
≥97.5%

Description

It is a source of cisoid butadiene for Diels-Alder reactions. This chemical is also a convenient source of sulfur dioxide. It is a solvent used in the petrochemical industry for the extraction of aromatics from hydrocarbon streams.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is a source of cisoid butadiene for Diels-Alder reactions. This chemical is also a convenient source of sulfur dioxide. It is a solvent used in the petrochemical industry for the extraction of aromatics from hydrocarbon streams.

Solubility
Soluble in water, ethanol, benzene, ether.

Notes
Store in a cool, dry, well-ventilated area. Store away from oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text