Thermo Scientific Chemicals

4-Hydroxy-3,5-dimethoxybenzyl alcohol, 97%, Thermo Scientific Chemicals

Catalog number: A13888.03
1 g, Each
Thermo Scientific Chemicals

4-Hydroxy-3,5-dimethoxybenzyl alcohol, 97%, Thermo Scientific Chemicals

Catalog number: A13888.03
1 g, Each
Quantity
Catalog number: A13888.03
also known as A13888-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
530-56-3
IUPAC Name
4-(hydroxymethyl)-2,6-dimethoxyphenol
Molecular Formula
C9H12O4
InChI Key
LUOAEJWSKPQLJD-UHFFFAOYSA-N
SMILES
COC1=CC(CO)=CC(OC)=C1O
Appearance (Color)
White to cream to yellow to pale brown
Melting Point (clear melt)
127.5-136.5?C
Assay (Silylated GC)
≥96.0%
Form
Powder
Identification (FTIR)
Conforms

Description

4-Hydroxy-3,5-dimethoxybenzyl alcohol is a 2,4,6- substituted phenol with antiviral properties.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Hydroxy-3,5-dimethoxybenzyl alcohol is a 2,4,6- substituted phenol with antiviral properties.

Solubility
Sparingly soluble (0.40 g/L at 25°C).

Notes
Store in a cool, dry, well-ventilated area. Store away from oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text