DL-Serine methyl ester hydrochloride, 98+%, Thermo Scientific Chemicals
DL-Serine methyl ester hydrochloride, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

DL-Serine methyl ester hydrochloride, 98+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
250 g
Catalog number A13904.09
also known as A13904-09
Price (USD)/ Each
29.10
-
Add to cart
Quantity:
10 g
Request bulk or custom format
Price (USD)/ Each
29.10
Add to cart
DL-Serine methyl ester hydrochloride, 98+%, Thermo Scientific Chemicals
Catalog numberA13904.09
Price (USD)/ Each
29.10
-
Add to cart
Chemical Identifiers
CAS5619-04-5
IUPAC Namehydrogen methyl 2-amino-3-hydroxypropanoate chloride
Molecular FormulaC4H10ClNO3
InChI KeyNDBQJIBNNUJNHA-UHFFFAOYNA-N
SMILES[H+].[Cl-].COC(=O)C(N)CO
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (Titration ex Chloride)≥98.0 to ≤102.0%
FormCrystals or powder or crystalline powder
Water Content (Karl Fischer Titration)≤1%
Appearance (Color)White to pale cream
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only