Tiglic acid, 97+%, Thermo Scientific Chemicals
Tiglic acid, 97+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Tiglic acid, 97+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number A13905.14
also known as A13905-14
Price (USD)/ Each
46.00
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
46.00
Add to cart
Tiglic acid, 97+%, Thermo Scientific Chemicals
Catalog numberA13905.14
Price (USD)/ Each
46.00
-
Add to cart
Chemical Identifiers
CAS80-59-1
IUPAC Name(2E)-2-methylbut-2-enoic acid
Molecular FormulaC5H8O2
InChI KeyUIERETOOQGIECD-ONEGZZNKSA-N
SMILESC\C=C(/C)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream to pale brown
FormCrystals or powder or crystalline powder
Assay (Aqueous acid-base Titration)≥97.0 to ≤103.0%
Assay (Silylated GC)≥97.0%
Melting Point (clear melt)60-67°C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only