Thermo Scientific Chemicals

3,4-Dichlorophenylhydrazine hydrochloride, 98+%, Thermo Scientific Chemicals

Catalog number: A13906.30
250 g, Each
Thermo Scientific Chemicals

3,4-Dichlorophenylhydrazine hydrochloride, 98+%, Thermo Scientific Chemicals

Catalog number: A13906.30
250 g, Each
Quantity
Catalog number: A13906.30
also known as A13906-30
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
19763-90-7
IUPAC Name
hydrogen (3,4-dichlorophenyl)hydrazine chloride
Molecular Formula
C6H7Cl3N2
InChI Key
WIKZRYSGQPZTMM-UHFFFAOYSA-N
SMILES
[H+].[Cl-].NNC1=CC=C(Cl)C(Cl)=C1
Appearance (Color)
White to cream to pale red
Assay (Titration ex Chloride)
≥98.0 to ≤102.0%
Form
Powder

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text