Thermo Scientific Chemicals

Ethyl 2-furoate, 99%, Thermo Scientific Chemicals

Catalog number: A13909.14
25 g, Each
Thermo Scientific Chemicals

Ethyl 2-furoate, 99%, Thermo Scientific Chemicals

Catalog number: A13909.14
25 g, Each
Quantity
Catalog number: A13909.14
also known as A13909-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
614-99-3
IUPAC Name
ethyl furan-2-carboxylate
Molecular Formula
C7H8O3
InChI Key
NHXSTXWKZVAVOQ-UHFFFAOYSA-N
SMILES
CCOC(=O)C1=CC=CO1
Appearance (Color)
White/colorless to cream or pale yellow to orange
Assay (GC)
≥98.5%
Melting Point (clear melt)
27-37?C
Form
Crystals or powder or crystalline powder or fused/lumpy solid

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text