3,5-Dichloroaniline, 98%, Thermo Scientific Chemicals
3,5-Dichloroaniline, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3,5-Dichloroaniline, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
50 g
Catalog number A13922.30
also known as A13922-30
Price (USD)/ Each
126.00
-
Add to cart
Quantity:
250 g
Request bulk or custom format
Price (USD)/ Each
126.00
Add to cart
3,5-Dichloroaniline, 98%, Thermo Scientific Chemicals
Catalog numberA13922.30
Price (USD)/ Each
126.00
-
Add to cart
Chemical Identifiers
CAS626-43-7
IUPAC Name3,5-dichloroaniline
Molecular FormulaC6H5Cl2N
InChI KeyUQRLKWGPEVNVHT-UHFFFAOYSA-N
SMILESNC1=CC(Cl)=CC(Cl)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals, crystalline powder, flakes or chunks
CommentMay darken on storage
Identification (FTIR)Conforms (non-U.S. specification)
Melting Point (clear melt)49-53°C
Appearance (Color)White to cream to brown
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only