Thermo Scientific Chemicals

1,10-Decanediol, 97%, Thermo Scientific Chemicals

Catalog number: A13923.22
100 g, Each
Thermo Scientific Chemicals

1,10-Decanediol, 97%, Thermo Scientific Chemicals

Catalog number: A13923.22
100 g, Each
Quantity
Catalog number: A13923.22
also known as A13923-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
112-47-0
IUPAC Name
decane-1,10-diol
Molecular Formula
C10H22O2
InChI Key
FOTKYAAJKYLFFN-UHFFFAOYSA-N
SMILES
OCCCCCCCCCCO
Form
Powder or flakes
Identification (FTIR)
Conforms
Appearance (Color)
White
Assay (GC)
≥96.0%

Description

1,10-Decanediol is an minor intermediate in the production of polyesters. It also used in essence, perfumes and in pharmaceuticals. The diacrylate is hydrophobic and used for flexible dental materials, radiation curable coatings, and for lenses.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,10-Decanediol is an minor intermediate in the production of polyesters. It also used in essence, perfumes and in pharmaceuticals. The diacrylate is hydrophobic and used for flexible dental materials, radiation curable coatings, and for lenses.

Solubility
Soluble in alcohol. Insoluble in water, petroleum ether.

Notes
Store in a cool, dry, well-ventilated area. Incompatible with oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text