Thermo Scientific Chemicals

2-Hydroxyphenylacetic acid, 98%, Thermo Scientific Chemicals

Catalog number: A13924.09
10 g, Each
Thermo Scientific Chemicals

2-Hydroxyphenylacetic acid, 98%, Thermo Scientific Chemicals

Catalog number: A13924.09
10 g, Each
Quantity
Catalog number: A13924.09
also known as A13924-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
614-75-5
IUPAC Name
2-(2-hydroxyphenyl)acetic acid
Molecular Formula
C8H8O3
InChI Key
CCVYRRGZDBSHFU-UHFFFAOYSA-N
SMILES
OC(=O)CC1=CC=CC=C1O
Assay (HPLC)
≥97.5% (U.K. Specification)
Assay from Supplier's CofA
≥97.5%, unspecified (U.S. Specification)
Comment
Purchased in U.K. and U.S.
Melting Point (clear melt)
141.0-147.0°C (U.K. Specification)
Appearance (Color)
Cream to yellow to pale brown

Description

2-Hydroxyphenylacetic Acid is an intermediate in the synthesis of atenolol and 3,4-dihydroxyphenylacetic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Hydroxyphenylacetic Acid is an intermediate in the synthesis of atenolol and 3,4-dihydroxyphenylacetic acid.

Solubility
Soluble in Methanol and Water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text