Thermo Scientific Chemicals

tert-Butyl hydroperoxide, 70% aq. soln., Thermo Scientific Chemicals

Catalog number: A13926.AE
100 mL, Each
Thermo Scientific Chemicals

tert-Butyl hydroperoxide, 70% aq. soln., Thermo Scientific Chemicals

Catalog number: A13926.AE
100 mL, Each
Quantity
Catalog number: A13926.AE
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
75-91-2
IUPAC Name
2-methylpropane-2-peroxol
Molecular Formula
C4H10O2
InChI Key
CIHOLLKRGTVIJN-UHFFFAOYSA-N
SMILES
CC(C)(C)OO
Appearance (Color)
Clear colorless
Available Oxygen
12.07-13.31 %
Form
Liquid
Peroxide content
68.0-73.0 %
Identification (FTIR)
Conforms

Description

tert-Butyl hydroperoxide is used as an initiator for radical polymerization and in various oxidation process such as sharpless epoxidation. It is involved in osmium catalyzed vicinal hydroxylation of olefins under alkaline conditions. Furthermore, it is used in catalytic asymmetric oxidation of sulfides to sulfoxides using binaphthol as a chiral auxiliary and in the oxidation of dibenzothiophenes. It plays an important role for the introduction of peroxy groups in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
tert-Butyl hydroperoxide is used as an initiator for radical polymerization and in various oxidation process such as sharpless epoxidation. It is involved in osmium catalyzed vicinal hydroxylation of olefins under alkaline conditions. Furthermore, it is used in catalytic asymmetric oxidation of sulfides to sulfoxides using binaphthol as a chiral auxiliary and in the oxidation of dibenzothiophenes. It plays an important role for the introduction of peroxy groups in organic synthesis.

Solubility
Miscible with water and diethyl ether.

Notes
Light sensitive. Store in cool place. Incompatible with powdered metals, strong oxidizing agents, reducing agents, acids, alkalis and heavy metals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text