Thermo Scientific Chemicals

Isopropyl 4-hydroxybenzoate, 98%, Thermo Scientific Chemicals

Catalog number: A13930.09
10 g, Each
Thermo Scientific Chemicals

Isopropyl 4-hydroxybenzoate, 98%, Thermo Scientific Chemicals

Catalog number: A13930.09
10 g, Each
Quantity
Catalog number: A13930.09
also known as A13930-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
4191-73-5
IUPAC Name
propan-2-yl 4-hydroxybenzoate
Molecular Formula
C10H12O3
InChI Key
CMHMMKSPYOOVGI-UHFFFAOYSA-N
SMILES
CC(C)OC(=O)C1=CC=C(O)C=C1
Form
Crystals or powder or crystalline powder
Melting Point
81.5-87.5?C
Identification (FTIR)
Conforms
Appearance (Color)
White to cream
Assay (GC)
≥97.5%

Description

It is used as a preservative in cosmetics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a preservative in cosmetics.

Solubility
Soluble in Chloroform, Ethyl Acetate.

Notes
Store in a cool, dry, well-ventilated area. Incompatible with oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text