Thermo Scientific Chemicals

Bromoacetonitrile, 96%, Thermo Scientific Chemicals

Catalog number: A13933.09
10 g, Each
Thermo Scientific Chemicals

Bromoacetonitrile, 96%, Thermo Scientific Chemicals

Catalog number: A13933.09
10 g, Each
Quantity
Catalog number: A13933.09
also known as A13933-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
590-17-0
IUPAC Name
2-bromoacetonitrile
Molecular Formula
C2H2BrN
InChI Key
REXUYBKPWIPONM-UHFFFAOYSA-N
SMILES
BrCC#N
Assay (GC)
≥95.0% (non-U.S. Specification)
Assay from Suppliers CofA
≥95.0% (U.S. specification)
Comment
Purchased in the U.S. and in other countries
Identification (FTIR)
Conforms (Non-U.S. specification)
Appearance (Color)
Clear colorless to yellow

Description

Bromoacetonitrile was used in the synthesis of 1-cyanomethyl-1,1-dimethylhydrazinium bromide. It is used as an alkalyting agent for the nitrogen phosphorus detector.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Bromoacetonitrile was used in the synthesis of 1-cyanomethyl-1,1-dimethylhydrazinium bromide. It is used as an alkalyting agent for the nitrogen phosphorus detector.

Solubility
Insoluble in water.

Notes
Store in a cool, dry, well-ventilated area. Incompatible with oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text