5-Fluoro-2-nitrotoluene, 98+%, Thermo Scientific Chemicals
5-Fluoro-2-nitrotoluene, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

5-Fluoro-2-nitrotoluene, 98+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number A13944.09
also known as A13944-09
Price (USD)/ Each
40.50
-
Add to cart
Quantity:
10 g
Request bulk or custom format
Price (USD)/ Each
40.50
Add to cart
5-Fluoro-2-nitrotoluene, 98+%, Thermo Scientific Chemicals
Catalog numberA13944.09
Price (USD)/ Each
40.50
-
Add to cart
Chemical Identifiers
CAS446-33-3
IUPAC Name4-fluoro-2-methyl-1-nitrobenzene
Molecular FormulaC7H6FNO2
InChI KeyJHFOWEGCZWLHNW-UHFFFAOYSA-N
SMILESCC1=CC(F)=CC=C1[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Refractive Index1.5260-1.5300 @ 20?C
Appearance (Color)Clear pale yellow to yellow
Assay (GC)≥98.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only