Thermo Scientific Chemicals

Cyclopropanemethylamine, 97%, Thermo Scientific Chemicals

Catalog number: A13946.03
1 g, Each
Thermo Scientific Chemicals

Cyclopropanemethylamine, 97%, Thermo Scientific Chemicals

Catalog number: A13946.03
1 g, Each
Quantity
Catalog number: A13946.03
also known as A13946-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2516-47-4
IUPAC Name
cyclopropylmethanaminium
Molecular Formula
C4H10N
InChI Key
IGSKHXTUVXSOMB-UHFFFAOYSA-O
SMILES
[NH3+]CC1CC1
Refractive Index
1.4300-1.4350 @ 20°C (non-U.S. specification)
Appearance (Color)
Clear colorless
Comment
Purchased in the U.S. and in other countries
Assay (GC)
≥96.0% (non-U.S. specification)
GC/MS Analysis
Conforms (U.S. specification)

Description

Cyclopropanemethylamine used in studies of potential inhibitors of glycine cleavage system of brain and liver mitochondria. It is used in chemical synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cyclopropanemethylamine used in studies of potential inhibitors of glycine cleavage system of brain and liver mitochondria. It is used in chemical synthesis.

Solubility
Fully miscible in water.

Notes
Air sensitive. Store in a cool, dry, well-ventilated area. Store away from oxidizing agent, air , carbon dioxide.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text