Thermo Scientific Chemicals

Potassium tert-butoxide, 97%, Thermo Scientific Chemicals

Catalog number: A13947.36
500 g, Each
Thermo Scientific Chemicals

Potassium tert-butoxide, 97%, Thermo Scientific Chemicals

Catalog number: A13947.36
500 g, Each
Quantity
Catalog number: A13947.36
also known as A13947-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
865-47-4
IUPAC Name
potassium 2-methylpropan-2-olate
Molecular Formula
C4H9KO
InChI Key
LPNYRYFBWFDTMA-UHFFFAOYSA-N
SMILES
[K+].CC(C)(C)[O-]
Appearance (Color)
White to pale cream
Form
Crystals or powder or crystalline powder
Assay (Aqueous acid-base Titration)
≥96.0 to ≤104.0% (UK-sourced material)
Assay from Supplier's CofA
≥96.0% (US-sourced material)
Comment
Material sourced in UK and US

Description

Potassium tert-butoxide is used as a strong non-nucleophilic base in organic chemistry. It plays an active role in dehydrohalogenation reactions. It is also useful for greener amidation of esters. It serves as an intermediate in Mizoroki-Heck-type reactions. Furthermore, it is used as an initiator in anionic polymerization of carbazolyl-substituted oxiranes. It catalyzes the reaction of hydrosilanes and heterocyclic compounds to give the silyl derivatives with evolution of hydrogen.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Potassium tert-butoxide is used as a strong non-nucleophilic base in organic chemistry. It plays an active role in dehydrohalogenation reactions. It is also useful for greener amidation of esters. It serves as an intermediate in Mizoroki-Heck-type reactions. Furthermore, it is used as an initiator in anionic polymerization of carbazolyl-substituted oxiranes. It catalyzes the reaction of hydrosilanes and heterocyclic compounds to give the silyl derivatives with evolution of hydrogen.

Solubility
Soluble in hexane, toluene, diethyl ether and terahydrofuran.

Notes
Air and moisture sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with water, acids, reducing agents, oxygen, alcohols, chlorinated solvents, halogens and ketones.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text