Thermo Scientific Chemicals

Ammonium carbonate, NH3 ca 30%, Thermo Scientific Chemicals

Catalog number: A13955.0B
1000 g, Each
Thermo Scientific Chemicals

Ammonium carbonate, NH3 ca 30%, Thermo Scientific Chemicals

Catalog number: A13955.0B
1000 g, Each
Quantity
Catalog number: A13955.0B
also known as A13955-0B
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
506-87-6
IUPAC Name
carbonic acid diamine
Molecular Formula
CH8N2O3
InChI Key
PRKQVKDSMLBJBJ-UHFFFAOYSA-N
SMILES
N.N.OC(O)=O
Form
Crystals or powder or crystalline powder or fused/lumpy solid or chunks
Assay from Supplier's CofA
28.0 - 34.0% as NH3
Appearance (Color)
White

Description

Ammonium carbonate is suitable as a leavening agent, pH-control agent, dough strengthener and texturizer. It may be used as a dispersant for SiO2 nano particles for dynamic light scattering, Z-potential, and centrifugal liquid sedimentation measurements and in the synthesis of ZnO nanoparticles. It may also be used as an eluent in the separation of silica nanoparticles by asymmetric flow field-flow fractionation (AF4) coupled to inductively coupled plasma mass . It was used in experiment to distinguish 26 different phenolic components and in Capillary electrophoresis-mass spectrometry (CE-MS/MS) as rapid screening technique.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ammonium carbonate is suitable as a leavening agent, pH-control agent, dough strengthener and texturizer. It may be used as a dispersant for SiO2 nano particles for dynamic light scattering, Z-potential, and centrifugal liquid sedimentation measurements and in the synthesis of ZnO nanoparticles. It may also be used as an eluent in the separation of silica nanoparticles by asymmetric flow field-flow fractionation (AF4) coupled to inductively coupled plasma mass . It was used in experiment to distinguish 26 different phenolic components and in Capillary electrophoresis-mass spectrometry (CE-MS/MS) as rapid screening technique.

Solubility
Soluble in water.

Notes
Moisture sensitive. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text