Thermo Scientific Chemicals

4-Formylphenoxyacetic acid, 98%, Thermo Scientific Chemicals

Catalog number: A13960.09
10 g, Each
Thermo Scientific Chemicals

4-Formylphenoxyacetic acid, 98%, Thermo Scientific Chemicals

Catalog number: A13960.09
10 g, Each
Quantity
Catalog number: A13960.09
also known as A13960-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
22042-71-3
Form
Powder
Appearance (Color)
Pale cream to cream to pale brown
Assay (Aqueous acid-base Titration)
≥97.5 to ≤102.5%
Melting Point (clear melt)
193.0-199.0?C
Assay (Silylated GC)
≥97.5%

Description

4-Formylphenoxyacetic acid is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Formylphenoxyacetic acid is used as an organic chemical synthesis intermediate.

Solubility
Soluble in hot methanol within almost transparency.

Notes
Air sensitive. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text