Thermo Scientific Chemicals

3-Methoxybenzaldehyde, 98%, Thermo Scientific Chemicals

Catalog number: A13962.36
500 g, Each
Thermo Scientific Chemicals

3-Methoxybenzaldehyde, 98%, Thermo Scientific Chemicals

Catalog number: A13962.36
500 g, Each
Quantity
Catalog number: A13962.36
also known as A13962-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
591-31-1
Free acid (titration)
≤1%
Appearance (Color)
Clear colorless to yellow
Assay (GC)
≥97.5%
Form
Liquid
Refractive Index
1.5515-1.5555 @ 20?C

Description

3-Methoxybenzaldehyde is used to prepare 3-(3-methoxy-phenyl)-1-phenyl-propenone by reaction with benzldehyde. It is used as an eluent for mono-13C isotopomers of vanillin in normal phase silica gel chromatography. It acts as an inhibitor of 4-(methylnitrosamino)-1-(3-pyridyl)-1-butanone (NNK) metabolism.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Methoxybenzaldehyde is used to prepare 3-(3-methoxy-phenyl)-1-phenyl-propenone by reaction with benzldehyde. It is used as an eluent for mono-13C isotopomers of vanillin in normal phase silica gel chromatography. It acts as an inhibitor of 4-(methylnitrosamino)-1-(3-pyridyl)-1-butanone (NNK) metabolism.

Solubility
Slightly soluble in water. Soluble in glacial acetic acid, alcohol, chloroform, ether, carbon disulfide, methanol, aqueous solutions of alkali hydroxides.

Notes
Air sensitive. Incompatible with strong bases, strong oxidizing agents and strong reducing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text