Thermo Scientific Chemicals

Suberic acid, 99%, Thermo Scientific Chemicals

Catalog number: A13963.0E
2500 g, Each
Thermo Scientific Chemicals

Suberic acid, 99%, Thermo Scientific Chemicals

Catalog number: A13963.0E
2500 g, Each
Quantity
Catalog number: A13963.0E
also known as A13963-0E
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
505-48-6
IUPAC Name
octanedioic acid
Molecular Formula
C8H14O4
InChI Key
TYFQFVWCELRYAO-UHFFFAOYSA-N
SMILES
OC(=O)CCCCCCC(O)=O
Form
Crystals or powder or crystalline powder
Assay (Aqueous acid-base Titration)
≥98.5 to ≤101.5%
Assay (Silylated GC)
≥98.5%
Identification (FTIR)
Conforms
Melting Point (clear melt)
139-146?C

Description

Suberic acid is used in drug synthesis and in the production of plastics. It is also used in the preparation of reduction-sensitive micelles, which affects their cellular uptake. Further, it is used in the delivery of anticancer drugs. In addition, it is used in the fluorescent detection of amidinium-carboxylate and amidinium formation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Suberic acid is used in drug synthesis and in the production of plastics. It is also used in the preparation of reduction-sensitive micelles, which affects their cellular uptake. Further, it is used in the delivery of anticancer drugs. In addition, it is used in the fluorescent detection of amidinium-carboxylate and amidinium formation.

Solubility
Soluble in water,dimethyl sulfoxide and methanol. Slightly soluble in ether. Insoluble in chloroform.

Notes
Incompatible with strong oxidizing agents, reducing agents and bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text