Thermo Scientific Chemicals

Methyl pyruvate, 98%, Thermo Scientific Chemicals

Catalog number: A13966.36
500 g, Each
Thermo Scientific Chemicals

Methyl pyruvate, 98%, Thermo Scientific Chemicals

Catalog number: A13966.36
500 g, Each
Quantity
Catalog number: A13966.36
also known as A13966-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
600-22-6
IUPAC Name
methyl 2-oxopropanoate
Molecular Formula
C4H6O3
InChI Key
CWKLZLBVOJRSOM-UHFFFAOYSA-N
SMILES
COC(=O)C(C)=O
Form
Liquid
Refractive Index
1.4020-1.4060 @ 20?C
Identification (FTIR)
Conforms
Appearance (Color)
Clear colorless to pale yellow to pale orange
Assay (GC)
≥97.5%

Description

Methyl Pyruvate acts as a potent secretagogue used in the study of stimulus-secretion coupling. Also used in the preparation of quinoxalines as dual phoosphodiesterase 2/10 (PDE2/10) inhibitors. It increases free calcium ion concentration in the cytosol of pancreatic cells. It is used in the preparation of dimethyl 2,3-dimethylenebutanedioate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl Pyruvate acts as a potent secretagogue used in the study of stimulus-secretion coupling. Also used in the preparation of quinoxalines as dual phoosphodiesterase 2/10 (PDE2/10) inhibitors. It increases free calcium ion concentration in the cytosol of pancreatic cells. It is used in the preparation of dimethyl 2,3-dimethylenebutanedioate.

Solubility
Soluble in chloroform, methanol, ether, alcohol. Slightly soluble in water.

Notes
Store away from oxidizing agent, heat.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text