Thermo Scientific Chemicals

Ethyl picolinate, 99%, Thermo Scientific Chemicals

Catalog number: A13973.18
50 g, Each
Thermo Scientific Chemicals

Ethyl picolinate, 99%, Thermo Scientific Chemicals

Catalog number: A13973.18
50 g, Each
Quantity
Catalog number: A13973.18
also known as A13973-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2524-52-9
IUPAC Name
ethyl pyridine-2-carboxylate
Molecular Formula
C8H9NO2
InChI Key
FQYYIPZPELSLDK-UHFFFAOYSA-N
SMILES
CCOC(=O)C1=CC=CC=N1
Form
Liquid
Appearance (Color)
Clear colorless to pale yellow
Assay (GC)
≥98.5%
Refractive Index
1.5090-1.5120 @ 20?C

Description

Ethyl picolinate is used in the preparation of 2-Aminodihydro[1,3]thiazines as BACE 2 inhibitors which is used in the treatment of diabetes. It is also used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl picolinate is used in the preparation of 2-Aminodihydro[1,3]thiazines as BACE 2 inhibitors which is used in the treatment of diabetes. It is also used as pharmaceutical intermediate.

Solubility
Fully miscible in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text