Thermo Scientific Chemicals

1,3,5-Trimethoxybenzene, 99%, Thermo Scientific Chemicals

Catalog number: A13981.09
10 g, Each
Thermo Scientific Chemicals

1,3,5-Trimethoxybenzene, 99%, Thermo Scientific Chemicals

Catalog number: A13981.09
10 g, Each
Quantity
Catalog number: A13981.09
also known as A13981-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
621-23-8
IUPAC Name
1,3,5-trimethoxybenzene
Molecular Formula
C9H12O3
InChI Key
LKUDPHPHKOZXCD-UHFFFAOYSA-N
SMILES
COC1=CC(OC)=CC(OC)=C1
Melting Point (clear melt)
50-54?C
Form
Crystals or powder or crystalline powder or lumps
Appearance (Color)
White to cream or pale yellow or pale pink
Identification (FTIR)
Conforms
Assay (GC)
≥98.5%

Description

1,3,5-Trimethoxybenzene is a key component of the Chinese rose odor. It is used as a cosmetic agents. It was employed as secondary standard in quantitative proton NMR spectroscopy of pharmaceuticals. It cleaves p-methoxybenzyl protecting group on various alcohols and acids

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,3,5-Trimethoxybenzene is a key component of the Chinese rose odor. It is used as a cosmetic agents. It was employed as secondary standard in quantitative proton NMR spectroscopy of pharmaceuticals. It cleaves p-methoxybenzyl protecting group on various alcohols and acids

Solubility
Insoluble in water.

Notes
Store in a cool, dry, well-ventilated area. Store away from oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text