Ethyl hexanoate, 99%, Thermo Scientific Chemicals
Ethyl hexanoate, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ethyl hexanoate, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
2500 g
100 g
500 g
Catalog number A13985.0E
also known as A13985-0E
Price (USD)/ Each
333.00
-
Add to cart
Quantity:
2500 g
Request bulk or custom format
Price (USD)/ Each
333.00
Add to cart
Ethyl hexanoate, 99%, Thermo Scientific Chemicals
Catalog numberA13985.0E
Price (USD)/ Each
333.00
-
Add to cart
Chemical Identifiers
CAS123-66-0
IUPAC Nameethyl hexanoate
Molecular FormulaC8H16O2
InChI KeySHZIWNPUGXLXDT-UHFFFAOYSA-N
SMILESCCCCCC(=O)OCC
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Refractive Index1.4060-1.4090 @ 20?C
Appearance (Color)Clear colorless
Assay (GC)≥98.5%
Identification (FTIR)Conforms
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only