Thermo Scientific™

D-Isosorbide, 98%, Thermo Scientific Chemicals

Catalog number: A13989.18
50 g, Each
Thermo Scientific™

D-Isosorbide, 98%, Thermo Scientific Chemicals

Catalog number: A13989.18
50 g, Each
Quantity
Catalog number: A13989.18
also known as A13989-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
652-67-5
IUPAC Name
(3R,3aR,6S,6aR)-hexahydrofuro[3,2-b]furan-3,6-diol
Molecular Formula
C6H10O4
InChI Key
KLDXJTOLSGUMSJ-JGWLITMVSA-N
SMILES
O[C@H]1CO[C@@H]2[C@H](O)CO[C@H]12
Appearance (Color)
White to cream to pale yellow
Water Content (Karl Fischer Titration)
≤1.5%
Form
Crystals or powder or crystalline powder or fused solid or flakes
Identification (FTIR)
Conforms
Assay (Silylated GC)
≥97.5%

Description

D-Isosorbide is used as a reagent in the preparation of detergents, agrochemicals, cleansers, cosmetics and vasodilators. It is also employed as a pharmaceutical intermediate for dimethyl ether and mono- and dinitrate derivatives, which is used as vasodilators and coronary disease therapeutics. It is used in the treatment of hydrocephalus and glaucoma. It finds application as a building block for polymers like polycarbonates and polyesters.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in alcohols, water and ketones.

Notes
Hygroscopic. Store in a cool place. Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text