D-Isosorbide, 98%, Thermo Scientific Chemicals
D-Isosorbide, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

D-Isosorbide, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
50 g
1000 g
Catalog number A13989.30
also known as A13989-30
Price (USD)/ Each
98.65
Online exclusive
109.00 
Save 10.35 (10%)
-
Add to cart
Quantity:
250 g
Request bulk or custom format
Price (USD)/ Each
98.65
Online exclusive
109.00 
Save 10.35 (10%)
Add to cart
D-Isosorbide, 98%, Thermo Scientific Chemicals
Catalog numberA13989.30
Price (USD)/ Each
98.65
Online exclusive
109.00 
Save 10.35 (10%)
-
Add to cart
Chemical Identifiers
CAS652-67-5
IUPAC Name(3R,3aR,6S,6aR)-hexahydrofuro[3,2-b]furan-3,6-diol
Molecular FormulaC6H10O4
InChI KeyKLDXJTOLSGUMSJ-JGWLITMVSA-N
SMILESO[C@H]1CO[C@@H]2[C@H](O)CO[C@H]12
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream to pale yellow
Water Content (Karl Fischer Titration)≤1.5%
FormCrystals or powder or crystalline powder or fused solid or flakes
Identification (FTIR)Conforms
Assay (Silylated GC)≥97.5%
View more
D-Isosorbide is used as a reagent in the preparation of detergents, agrochemicals, cleansers, cosmetics and vasodilators. It is also employed as a pharmaceutical intermediate for dimethyl ether and mono- and dinitrate derivatives, which is used as vasodilators and coronary disease therapeutics. It is used in the treatment of hydrocephalus and glaucoma. It finds application as a building block for polymers like polycarbonates and polyesters.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in alcohols, water and ketones.

Notes
Hygroscopic. Store in a cool place. Incompatible with strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Parikh, K.; Mistry, B.; Jana, S.; Gajaria, T.; Gupta, S.; Devkar, R. V.; Kumar, S. Isosorbide spacer containing gemini surfactants: surface and biochemical study. Colloid. Polym. Sci. 2015, 293 (5), 1437-1446.
  2. Lavilla, C.; Munoz-Guerra, S. Sugar-based aromatic copolyesters: a comparative study regarding isosorbide and diacetalized alditols as sustainable comonomers. Green Chem. 2013, 15 (1), 144-151.