Thermo Scientific Chemicals

3-Methyl-2-cyclopenten-1-one, 98%, stab. with 0.1% hydroquinone, Thermo Scientific Chemicals

Catalog number: A13993.09
10 g, Each
Thermo Scientific Chemicals

3-Methyl-2-cyclopenten-1-one, 98%, stab. with 0.1% hydroquinone, Thermo Scientific Chemicals

Catalog number: A13993.09
10 g, Each
Quantity
Catalog number: A13993.09
also known as A13993-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2758-18-1
IUPAC Name
3-methylcyclopent-2-en-1-one
Molecular Formula
C6H8O
InChI Key
CHCCBPDEADMNCI-UHFFFAOYSA-N
SMILES
CC1=CC(=O)CC1
Form
Liquid
Refractive Index
1.4870-1.4910 @ 20?C
Appearance (Color)
Clear, colorless to yellow
Assay (GC)
>97.5%

Description

It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals.

Solubility
Fully miscible in water.

Notes
Store in a cool, dry, well-ventilated area. Store away from oxidizing agent, heat. Store in refrigerator.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text