3-Methyl-2-cyclopenten-1-one, 98%, stab. with 0.1% hydroquinone, Thermo Scientific Chemicals
3-Methyl-2-cyclopenten-1-one, 98%, stab. with 0.1% hydroquinone, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-Methyl-2-cyclopenten-1-one, 98%, stab. with 0.1% hydroquinone, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
10 g
Catalog number A13993.18
also known as A13993-18
Price (USD)/ Each
266.65
Online exclusive
296.00 
Save 29.35 (10%)
-
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (USD)/ Each
266.65
Online exclusive
296.00 
Save 29.35 (10%)
Add to cart
3-Methyl-2-cyclopenten-1-one, 98%, stab. with 0.1% hydroquinone, Thermo Scientific Chemicals
Catalog numberA13993.18
Price (USD)/ Each
266.65
Online exclusive
296.00 
Save 29.35 (10%)
-
Add to cart
Chemical Identifiers
CAS2758-18-1
IUPAC Name3-methylcyclopent-2-en-1-one
Molecular FormulaC6H8O
InChI KeyCHCCBPDEADMNCI-UHFFFAOYSA-N
SMILESCC1=CC(=O)CC1
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Refractive Index1.4870-1.4910 @ 20?C
Appearance (Color)Clear, colorless to yellow
Assay (GC)>97.5%
It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals.

Solubility
Fully miscible in water.

Notes
Store in a cool, dry, well-ventilated area. Store away from oxidizing agent, heat. Store in refrigerator.
RUO – Research Use Only