4-tert-Butylbenzoic acid as a thermal stabilizer in PVC. It is used as a intermediate in chemical industry for producing esters of PTBBA, a chain stop agent in resins. It used as a potent yeast sirtuin (Sir2p) inhibitor.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-tert-Butylbenzoic acid as a thermal stabilizer in PVC. It is used as a intermediate in chemical industry for producing esters of PTBBA, a chain stop agent in resins. It used as a potent yeast sirtuin (Sir2p) inhibitor.
Solubility
Insoluble in water.
Notes
Store in a cool, dry, well-ventilated area. Store away from oxidizing agent.
RUO – Research Use Only
Yi-Pei Chen; Chad C Catbagan; Jeannette T Bowler; Trevor Gokey; Natalie D M Goodwin; Anton B Guliaev; Weiming Wu; Taro Amagata. Evaluation of benzoic acid derivatives as sirtuin inhibitors. Bioorganic & Medicinal Chemistry Letters. 2014, 24 (1), 349-352.
R Carabias-Martínez; E Rodríguez-Gonzalo; P Revilla-Ruiz. Determination of weakly acidic endocrine-disrupting compounds by liquid chromatography-mass spectrometry with post-column base addition. Journal of Chromatography. 2004, 1056 (1-2), 131-138.