Thermo Scientific Chemicals

1,4-Diazabicyclo[2.2.2]octane, 98%, Thermo Scientific Chemicals

Catalog number: A14003.36
500 g, Each
Thermo Scientific Chemicals

1,4-Diazabicyclo[2.2.2]octane, 98%, Thermo Scientific Chemicals

Catalog number: A14003.36
500 g, Each
Quantity
Catalog number: A14003.36
also known as A14003-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
280-57-9
IUPAC Name
1,4-diazabicyclo[2.2.2]octane
Molecular Formula
C6H12N2
InChI Key
IMNIMPAHZVJRPE-UHFFFAOYSA-N
SMILES
C1CN2CCN1CC2
Assay (Non-aqueous acid-base Titration)
≥97.5 to ≤102.5% (non-U.S. sourced material)
Appearance (Color)
White to cream
Assay from Supplier's CofA
≥97.5% (GC, U.S. sourced material)
Identification (FTIR)
Conforms (non-U.S. sourced material)
Melting Point (clear melt)
153-162°C (non-U.S. sourced material)

Description

1,4-Diazabicyclo[2.2.2]octane is used as polyurethane catalyst, Balis-Hillman reaction catalyst complexing ligand and lewis base. It finds use in dye lasers and in mounting samples for fluorescence microscopy and as anti-fade reagent shown to scavenge free radicals due to flurochrome excitation of fluorochromes. Further, it is an oxidation and polymerization catalyst.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,4-Diazabicyclo[2.2.2]octane is used as polyurethane catalyst, Balis-Hillman reaction catalyst complexing ligand and lewis base. It finds use in dye lasers and in mounting samples for fluorescence microscopy and as anti-fade reagent shown to scavenge free radicals due to flurochrome excitation of fluorochromes. Further, it is an oxidation and polymerization catalyst.

Solubility
Soluble in water, acetone, benzene, ethanol and methyl ethyl ketone.

Notes
Hygroscopic. Incompatible with strong oxidizing agents and strong acids.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text