Dimethyl phenylphosphonite, 98%, Thermo Scientific Chemicals
Dimethyl phenylphosphonite, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Dimethyl phenylphosphonite, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number A14006.03
also known as A14006-03
Price (USD)/ Each
27.20
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
27.20
Add to cart
Dimethyl phenylphosphonite, 98%, Thermo Scientific Chemicals
Catalog numberA14006.03
Price (USD)/ Each
27.20
-
Add to cart
Chemical Identifiers
CAS2946-61-4
IUPAC Namedimethyl phenylphosphonite
Molecular FormulaC8H11O2P
InChI KeyLMZLQYYLELWCCW-UHFFFAOYSA-N
SMILESCOP(OC)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
FormLiquid
Assay (GC)≥97.5%
Refractive Index1.5265-1.5305 @ 20?C
Dimethyl phenylphosphonite is used in the preparation of phenyl-phosphonic acid dimethyl ester. It is used as a precursor for the preparation of dinitrosyltris(dimethyl phenylphosphonite)manganese(I) tetrafluoroborate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dimethyl phenylphosphonite is used in the preparation of phenyl-phosphonic acid dimethyl ester. It is used as a precursor for the preparation of dinitrosyltris(dimethyl phenylphosphonite)manganese(I) tetrafluoroborate.

Solubility
Miscible with alcohol. Immiscible with water.

Notes
Store in a cool place. Incompatible with strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Mai, S.; Xu, M.; Liao, X.; Xing, L.; Li, W. Improving cyclic stability of lithium nickel manganese oxide cathode at elevated temperature by using dimethyl phenylphosphonite as electrolyte additive. J. Power Sources 2015, 273, 816-822.
  2. Elomaa, L.; Pan, C. C.; Shanjani, Y.; Malkovskiy, A.; Seppälä, J. V.; Yang, Y. Three-dimensional fabrication of cell-laden biodegradable poly(ethylene glycol-co-depsipeptide) hydrogels by visible light stereolithography. J. Mater. Chem. B 2015, 3 (42), 8348-8358.