Thermo Scientific Chemicals

Dimethyl phenylphosphonite, 98%, Thermo Scientific Chemicals

Catalog number: A14006.06
5 g, Each
Thermo Scientific Chemicals

Dimethyl phenylphosphonite, 98%, Thermo Scientific Chemicals

Catalog number: A14006.06
5 g, Each
Quantity
Catalog number: A14006.06
also known as A14006-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2946-61-4
IUPAC Name
dimethyl phenylphosphonite
Molecular Formula
C8H11O2P
InChI Key
LMZLQYYLELWCCW-UHFFFAOYSA-N
SMILES
COP(OC)C1=CC=CC=C1
Appearance (Color)
Clear colorless
Form
Liquid
Assay (GC)
≥97.5%
Refractive Index
1.5265-1.5305 @ 20?C

Description

Dimethyl phenylphosphonite is used in the preparation of phenyl-phosphonic acid dimethyl ester. It is used as a precursor for the preparation of dinitrosyltris(dimethyl phenylphosphonite)manganese(I) tetrafluoroborate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dimethyl phenylphosphonite is used in the preparation of phenyl-phosphonic acid dimethyl ester. It is used as a precursor for the preparation of dinitrosyltris(dimethyl phenylphosphonite)manganese(I) tetrafluoroborate.

Solubility
Miscible with alcohol. Immiscible with water.

Notes
Store in a cool place. Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text