3-Picoline, 99%, Thermo Scientific Chemicals
3-Picoline, 99%, Thermo Scientific Chemicals
Thermo Scientific™

3-Picoline, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
2500 mL
100 mL
Catalog number A14012.0F
also known as A14012-0F
Price (USD)/ Each
145.00
-
Add to cart
Quantity:
2500 mL
Request bulk or custom format
Price (USD)/ Each
145.00
Add to cart
3-Picoline, 99%, Thermo Scientific Chemicals
Catalog numberA14012.0F
Price (USD)/ Each
145.00
-
Add to cart
Chemical Identifiers
CAS108-99-6
IUPAC Name3-methylpyridine
Molecular FormulaC6H7N
InChI KeyITQTTZVARXURQS-UHFFFAOYSA-N
SMILESCC1=CC=CN=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Water Content≤0.5%
Appearance (Color)Clear colorless to yellow
Assay from Supplier's CofA≥98.5%
3-Picoline is used as a precursor in pharmaceuticals and agricultural industries. It acts as a precursor to 3-cyanopyridine, niacin, vitamin-B. It is an antidote for organophosphate poisoning.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Picoline is used as a precursor in pharmaceuticals and agricultural industries. It acts as a precursor to 3-cyanopyridine, niacin, vitamin-B. It is an antidote for organophosphate poisoning.

Solubility
Miscible with water, alcohol, ether, acetone, carbon tetrachloride.

Notes
Hygroscopic. Incompatible with acids, acid chlorides and chloroformates.
RUO – Research Use Only

General References:

  1. Du, Q.; Wang, T.; Wang, Z.; Jiang, X.; Wang, L. Rapid Determination of Glutamine in Human Plasma by High-Performance Liquid Chromatographic-Tandem Mass Spectrometry and Its Application in Pharmacokinetic Studies. J. Chromatogr. Sci. 2015, 53 (1), 79-84.
  2. Cheng, C.; Hartwig, J. F. Iridium-Catalyzed Silylation of Aryl C-H Bonds. J. Am. Chem. Soc. 2015, 137 (2), 592-595.