Thermo Scientific Chemicals

3,3-Dimethyl-1-butyne, 98%, Thermo Scientific Chemicals

Catalog number: A14013.06
5 g, Each
Thermo Scientific Chemicals

3,3-Dimethyl-1-butyne, 98%, Thermo Scientific Chemicals

Catalog number: A14013.06
5 g, Each
Quantity
Catalog number: A14013.06
also known as A14013-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
917-92-0
IUPAC Name
3,3-dimethylbut-1-yne
Molecular Formula
C6H10
InChI Key
PPWNCLVNXGCGAF-UHFFFAOYSA-N
SMILES
CC(C)(C)C#C
Assay (GC)
≥97.5%
Appearance (Color)
Clear colorless to pale yellow
Form
Liquid

Description

3,3-Dimethyl-1-butyne is used as an active pharmaceutical intermediate for terbinafine. It is also used in the preparation of 2,2,7,7-tetramethyl-octa-3,5-diyne. Further, it is used in the synthesis of erythro and threo isomers of B-(3,3-dimethyl-1,2-dideuterio-1-butyl)-9-BBN by hydroboration-deuteronolysis-hydroboration sequence.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,3-Dimethyl-1-butyne is used as an active pharmaceutical intermediate for terbinafine. It is also used in the preparation of 2,2,7,7-tetramethyl-octa-3,5-diyne. Further, it is used in the synthesis of erythro and threo isomers of B-(3,3-dimethyl-1,2-dideuterio-1-butyl)-9-BBN by hydroboration-deuteronolysis-hydroboration sequence.

Solubility
Miscible with chloroform, benzene and toluene. Immiscible with water.

Notes
Store in a cool place. Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text