Thermo Scientific Chemicals

2-Methoxybenzyl alcohol, 99%, Thermo Scientific Chemicals

Catalog number: A14016.14
25 g, Each
Thermo Scientific Chemicals

2-Methoxybenzyl alcohol, 99%, Thermo Scientific Chemicals

Catalog number: A14016.14
25 g, Each
Quantity
Catalog number: A14016.14
also known as A14016-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
612-16-8
IUPAC Name
(2-methoxyphenyl)methanol
Molecular Formula
C8H10O2
InChI Key
WYLYBQSHRJMURN-UHFFFAOYSA-N
SMILES
COC1=CC=CC=C1CO
Assay (GC)
≥98.5%
Identification (FTIR)
Conforms
Refractive Index
1.5460-1.5490 @ 20?C
Form
Liquid
Appearance (Color)
Clear colorless

Description

It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals.

Solubility
Not miscible or difficult to mix in water. Soluble in ethanol.

Notes
Store in a cool, dry, well-ventilated area. Store away from oxidizing agent, strong acids.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text