Thermo Scientific Chemicals

3-Bromothiophene, 97%, Thermo Scientific Chemicals

Catalog number: A14022.18
50 g, Each
Thermo Scientific Chemicals

3-Bromothiophene, 97%, Thermo Scientific Chemicals

Catalog number: A14022.18
50 g, Each
Quantity
Catalog number: A14022.18
also known as A14022-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
872-31-1
IUPAC Name
3-bromothiophene
Molecular Formula
C4H3BrS
InChI Key
XCMISAPCWHTVNG-UHFFFAOYSA-N
SMILES
BrC1=CSC=C1
Form
Liquid
Refractive Index
1.5895-1.5945 @ 20?C
Assay (GC)
≥96.0%
Appearance (Color)
Clear colorless to pale yellow
Identification (FTIR)
Conforms

Description

3-Bromothiophene is used in the preparation of derivatives such as thienylenic alpha, μ-diformyl-alpha-oligothiophenes and 3-lithiothiophene. It is also used in the synthesis of 3-lithiothiophene by reacting with n-butyllithium.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Bromothiophene is used in the preparation of derivatives such as thienylenic alpha, µ-diformyl-alpha-oligothiophenes and 3-lithiothiophene. It is also used in the synthesis of 3-lithiothiophene by reacting with n-butyllithium.

Solubility
Immiscible with water.

Notes
Light sensitive. Incompatible with strong oxidizing agents, strong reducing agents and strong bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text