Glycerol monoacetate is used for production of tanning leather and dye. It is also used as a solvent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Glycerol monoacetate is used for production of tanning leather and dye. It is also used as a solvent.
Solubility
Soluble in water, methanol, ethanol, ether (slightly), and insoluble in benzene.
Notes
Hygroscopic. Protect from moisture. Store at room temperature. Store away from strong oxidizing agents.
RUO – Research Use Only
R.A. Peters; P.L. Morselli. Observations on the use and action of monoacetin in fluoroacetate poisoning. Biochemical Pharmacology. 1965, 14 (12), 1891-1893.
Takuya Fukumura; Takuji Toda; Yuichiro Seki; Masaki Kubo; Naomi Shibasaki-Kitakawa and Toshikuni Yonemoto. Catalytic Synthesis of Glycerol Monoacetate Using a Continuous Expanded Bed Column Reactor Packed with Cation-Exchange Resin. Ind. Eng. Chem. Res. 2009, 48 (4), 1816-1823.