Thermo Scientific Chemicals

1-Bromoheptane, 98+%, Thermo Scientific Chemicals

Catalog number: A14031.0B
1000 g, Each
Thermo Scientific Chemicals

1-Bromoheptane, 98+%, Thermo Scientific Chemicals

Catalog number: A14031.0B
1000 g, Each
Quantity
Catalog number: A14031.0B
also known as A14031-0B
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
629-04-9
IUPAC Name
1-bromoheptane
Molecular Formula
C7H15Br
InChI Key
LSXKDWGTSHCFPP-UHFFFAOYSA-N
SMILES
CCCCCCCBr
Appearance (Color)
Clear colorless to pale yellow
Form
Liquid
Assay (GC)
≥98.0%
Identification (FTIR)
Conforms
Refractive Index
1.4485-1.4525 @ 20?C

Description

1-bromoheptane is an important precursor material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-bromoheptane is an important precursor material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.

Solubility
Immiscible with water.

Notes
Flammable and incompatible with oxidizing agents, strong bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text