1-bromoheptane is an important precursor material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1-bromoheptane is an important precursor material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.
Solubility
Immiscible with water.
Notes
Flammable and incompatible with oxidizing agents, strong bases.
RUO – Research Use Only
General References:
- Qu, H.; Li, H.; Liang, F.; Zhang, B. The 1-bromoheptane photodissociation near 234 nm. Chem. Phys. 2006, 330 (3), 355-359.
- Sun, C.; Clarke, S. M.; Brewer, A.; Li, B.; Parker, J. E.; Demmel, F. The structures of 1-bromoheptane and 1-bromononane monolayers adsorbed on the surface of graphite. Mol. Phys. 2012, 110 (4), 217-225.