Thermo Scientific Chemicals

Benzyltriphenylphosphonium chloride, 99%, Thermo Scientific Chemicals

Catalog number: A14034.36
500 g, Each
Thermo Scientific Chemicals

Benzyltriphenylphosphonium chloride, 99%, Thermo Scientific Chemicals

Catalog number: A14034.36
500 g, Each
Quantity
Catalog number: A14034.36
also known as A14034-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1100-88-5
IUPAC Name
benzyltriphenylphosphanium chloride
Molecular Formula
C25H22ClP
InChI Key
USFRYJRPHFMVBZ-UHFFFAOYSA-M
SMILES
[Cl-].C(C1=CC=CC=C1)[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
Appearance (Color)
White
Form
Powder
Assay (Titration ex Chloride)
≥98.5 to ≤101.5%
Water Content (Karl Fischer Titration)
≤0.5%
Melting Point
>300?C

Description

Benzyltriphenylphosphonium chloride is used as the accelerator of the Bisphenol AF curing system for fluoroelastomer. Also used as phase-transfer catalysts in the synthesis of certain organic compounds. It is used as a reactant for the synthesis of chemical compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Benzyltriphenylphosphonium chloride is used as the accelerator of the Bisphenol AF curing system for fluoroelastomer. Also used as phase-transfer catalysts in the synthesis of certain organic compounds. It is used as a reactant for the synthesis of chemical compounds.

Solubility
Soluble in water and alcohol. Insoluble in acetone.

Notes
Hygroscopic. Store under inert gas. Store away from strong oxidizing agents, water/ moisture..
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text